Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689440.3 | XP_005689497.2 | 4 - 5 | 448 | 112 | |
XM_005689441.3 | XP_005689498.2 | 4 - 5 | 407 | 112 | ||
XM_005689442.3 | XP_005689499.2 | 5 - 6 | 418 | 112 | ||
XM_005689443.3 | XP_005689500.2 | 4 - 5 | 958 | 112 | ||
XM_005689444.3 | XP_005689501.2 | 4 - 5 | 411 | 112 | ||
GenBank mRNA | JO424465.1 | - | 2 - 3 | 1753 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |