Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692115.3 | XP_005692172.2 | 3 - 4 | 476 | 84 | |
GenBank mRNA | JO419999.2 | - | 3 - 4 | 540 | 84 | |
JO431746.2 | - | 3 - 4 | 497 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |