Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063664.1 | XP_017919153.1 | 3 - 4 | 1061 | 82 | |
XM_018063665.1 | XP_017919154.1 | 3 - 4 | 1065 | 82 | ||
XM_018063666.1 | XP_017919155.1 | 3 - 4 | 402 | 82 | ||
GenBank mRNA | JO412193.2 | - | 3 - 4 | 512 | 82 | |
KA306124.1 | - | 1 - 2 | 2206 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |