Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698466.3 | XP_005698523.1 | 6 - 7 | 1598 | 91 | |
XM_005698467.3 | XP_005698524.1 | 6 - 7 | 1589 | 91 | ||
XM_018041275.1 | XP_017896764.1 | 6 - 7 | 1584 | 91 | ||
XM_018041276.1 | XP_017896765.1 | 6 - 7 | 1687 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |