Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXSM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042130.1 | XP_017897619.1 | 2 - 3 | 1184 | 86 | |
XM_018042131.1 | XP_017897620.1 | 2 - 3 | 1069 | 86 | ||
GenBank mRNA | JO371604.2 | - | 2 - 3 | 1179 | 86 | |
JT692515.1 | - | 2 - 3 | 1030 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |