Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056864.1 | XP_017912353.1 | 1 - 2 | 398 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |