Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057341.1 | XP_017912830.1 | 5 - 6 | 941 | 95 | |
XM_018057348.1 | XP_017912837.1 | 4 - 5 | 842 | 95 | ||
XM_018057357.1 | XP_017912846.1 | 3 - 4 | 437 | 95 | ||
GenBank mRNA | KA324102.1 | - | 4 - 5 | 558 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |