Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CANT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063667.1 | XP_017919156.1 | 2 - 3 | 746 | 73 | |
XM_018063668.1 | XP_017919157.1 | 3 - 4 | 867 | 73 | ||
XM_018063669.1 | XP_017919158.1 | 3 - 4 | 842 | 73 | ||
GenBank mRNA | JO411353.2 | - | 2 - 3 | 738 | 73 | |
JO516130.2 | - | 1 - 2 | 182 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |