Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676652.3 | XP_005676709.1 | 2 - 3 | 553 | 98 | |
XM_005676653.3 | XP_005676710.1 | 2 - 3 | 479 | 98 | ||
XM_018058549.1 | XP_017914038.1 | 2 - 3 | 654 | 98 | ||
GenBank mRNA | JO426679.1 | - | 2 - 3 | 732 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |