Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676652.3 | XP_005676709.1 | 4 - 5 | 1684 | 105 | |
XM_005676653.3 | XP_005676710.1 | 4 - 5 | 1610 | 105 | ||
XM_018058549.1 | XP_017914038.1 | 4 - 5 | 1785 | 105 | ||
GenBank mRNA | JO426679.1 | - | 4 - 5 | 1863 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |