Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678357.3 | XP_005678414.1 | 4 - 5 | 1212 | 70 | |
GenBank mRNA | KA352846.1 | - | 2 - 3 | 471 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |