Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058104.1 | XP_017913593.1 | 13 - 14 | 1298 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |