Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694858.3 | XP_005694915.1 | 3 - 4 | 393 | 79 | |
GenBank mRNA | JO444597.2 | - | 3 - 4 | 527 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |