Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696417.3 | XP_005696474.2 | 4 - 5 | 609 | 91 | |
XM_005696421.3 | XP_005696478.2 | 3 - 4 | 586 | 91 | ||
XM_013973902.2 | XP_013829356.2 | 5 - 6 | 927 | 91 | ||
XM_013973903.2 | XP_013829357.2 | 5 - 6 | 989 | 91 | ||
XM_013973904.2 | XP_013829358.2 | 5 - 6 | 632 | 91 | ||
XM_013973906.2 | XP_013829360.2 | 3 - 4 | 399 | 91 | ||
XM_018039201.1 | XP_017894690.1 | 5 - 6 | 1006 | 91 | ||
XM_018039202.1 | XP_017894691.1 | 5 - 6 | 770 | 91 | ||
XM_018039203.1 | XP_017894692.1 | 5 - 6 | 892 | 91 | ||
XM_018039204.1 | XP_017894693.1 | 5 - 6 | 761 | 91 | ||
XM_018039205.1 | XP_017894694.1 | 4 - 5 | 632 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons |