Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058774.1 | XP_017914263.1 | 2 - 3 | 1311 | 93 | |
XM_018058775.1 | XP_017914264.1 | 2 - 3 | 1290 | 93 | ||
XM_018058776.1 | XP_017914265.1 | 2 - 3 | 1311 | 93 | ||
XM_018058777.1 | XP_017914266.1 | 2 - 3 | 1290 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |