Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061516.1 | XP_017917005.1 | 11 - 12 | 12410 | 72 | |
GenBank mRNA | KA323498.1 | - | 3 - 4 | 1692 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |