Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674719.3 | XP_005674776.1 | 4 - 5 | 1241 | 63 | |
GenBank mRNA | JO356002.2 | - | 4 - 5 | 555 | 63 | |
JO395592.2 | - | 1 - 2 | 305 | 63 | ||
JO395593.2 | - | 1 - 2 | 305 | 63 | ||
JO415037.2 | - | 2 - 3 | 466 | 63 | ||
JO434715.2 | - | 2 - 3 | 725 | 63 | ||
JO523644.2 | - | 2 - 3 | 148 | 63 | ||
JO540338.2 | - | 4 - 5 | 629 | 63 | ||
JO586011.2 | - | 4 - 5 | 622 | 63 | ||
JT658637.1 | - | 4 - 5 | 561 | 63 | ||
JT705973.1 | - | 4 - 5 | 597 | 63 | ||
JT717630.1 | - | 4 - 5 | 602 | 63 | ||
JT738017.1 | - | 4 - 5 | 488 | 63 | ||
KA342963.1 | - | 4 - 5 | 739 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |