Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6H4orf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681496.3 | XP_005681553.2 | 3 - 4 | 198 | 89 | |
XM_018049362.1 | XP_017904851.1 | 3 - 4 | 214 | 89 | ||
GenBank mRNA | JO403741.2 | - | 1 - 2 | 495 | 89 | |
JO430518.2 | - | 1 - 2 | 1789 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |