Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PM20D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684818.3 | XP_005684875.3 | 6 - 7 | 1249 | 104 | |
GenBank mRNA | JO408772.2 | - | 1 - 2 | 172 | 104 | |
JO426190.1 | - | 7 - 8 | 1252 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |