Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18H16orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692120.3 | XP_005692177.1 | 14 - 15 | 1191 | 86 | |
XM_005692121.3 | XP_005692178.1 | 14 - 15 | 1177 | 86 | ||
XM_018062103.1 | XP_017917592.1 | 14 - 15 | 1179 | 86 | ||
GenBank mRNA | JT678375.1 | - | 15 - 16 | 1259 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |