Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18H16orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692120.3 | XP_005692177.1 | 8 - 9 | 684 | 70 | |
XM_005692121.3 | XP_005692178.1 | 8 - 9 | 670 | 70 | ||
XM_018062103.1 | XP_017917592.1 | 8 - 9 | 672 | 70 | ||
GenBank mRNA | JO587830.2 | - | 3 - 4 | 265 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |