Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043676.1 | XP_017899165.1 | 577 | 68 |
Gene Symbol: LOC108634359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044029.1 | XP_017899518.1 | 1 | 200 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Both primers and probe map within a single exon |