Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048932.1 | XP_017904421.1 | 3 - 4 | 626 | 95 | |
GenBank mRNA | JO337979.2 | - | 3 - 4 | 403 | 95 | |
JO640929.2 | - | 3 - 4 | 738 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |