Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048932.1 | XP_017904421.1 | 6 - 7 | 1152 | 76 | |
GenBank mRNA | JO640929.2 | - | 6 - 7 | 1264 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |