Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051858.1 | XP_017907347.1 | 6 - 7 | 3228 | 68 | |
XM_018051859.1 | XP_017907348.1 | 7 - 8 | 3342 | 68 | ||
XM_018051860.1 | XP_017907349.1 | 6 - 7 | 3235 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |