Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064726.1 | XP_017920215.1 | 3 - 4 | 252 | 78 | |
GenBank mRNA | JO447109.2 | - | 3 - 4 | 224 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |