Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041179.1 | XP_017896668.1 | 4 - 5 | 1312 | 81 | |
XM_018041180.1 | XP_017896669.1 | 4 - 5 | 1164 | 81 | ||
XM_018041181.1 | XP_017896670.1 | 4 - 5 | 1149 | 81 | ||
GenBank mRNA | JO502749.2 | - | 3 - 4 | 960 | 81 | |
JO631291.2 | - | 1 - 2 | 1032 | 81 | ||
KA306350.1 | - | 1 - 2 | 3655 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |