Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHNAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042689.1 | XP_017898178.1 | 5 - 6 | 736 | 102 | |
GenBank mRNA | KA347340.1 | - | 5 - 6 | 709 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |