Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684032.2 | XP_005684089.1 | 5 - 6 | 567 | 94 | |
XM_005684034.2 | XP_005684091.1 | 4 - 5 | 492 | 94 | ||
XM_005684035.2 | XP_005684092.1 | 4 - 5 | 486 | 94 | ||
GenBank mRNA | JT763177.1 | - | 2 - 3 | 998 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |