Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMAC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685634.3 | XP_005685691.1 | 2 - 3 | 197 | 76 | |
GenBank mRNA | JO380742.2 | - | 3 - 4 | 557 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |