Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB21L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691180.1 | XP_005691237.1 | 1 | 2121 | 104 | |
GenBank mRNA | JO643804.2 | - | 1 | 486 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |