Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693262.3 | XP_005693319.1 | 4 - 5 | 847 | 85 | |
XM_005693263.3 | XP_005693320.1 | 3 - 4 | 760 | 85 | ||
XM_005693264.3 | XP_005693321.1 | 3 - 4 | 597 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |