Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYATL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696397.3 | XP_005696454.2 | 4 - 5 | 455 | 71 | |
GenBank mRNA | JO397937.2 | - | 4 - 5 | 461 | 71 |
Gene Symbol: LOC108635060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045363.1 | XP_017900852.1 | 4 - 5 | 442 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |