Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049045.1 | XP_017904534.1 | 8 - 9 | 1017 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |