Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059233.1 | XP_017914722.1 | 4 - 5 | 400 | 109 | |
GenBank mRNA | JO636451.2 | - | 2 - 3 | 911 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |