Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973489.2 | XP_013828943.1 | 1 - 2 | 90 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |