Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040963.1 | XP_017896452.1 | 2 - 3 | 1015 | 110 | |
XM_018040964.1 | XP_017896453.1 | 2 - 3 | 1047 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |