Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676002.3 | XP_005676059.1 | 3 - 4 | 902 | 76 | |
XM_005676003.3 | XP_005676060.1 | 3 - 4 | 699 | 76 | ||
XM_013969089.2 | XP_013824543.1 | 3 - 4 | 706 | 76 | ||
XM_013969092.2 | XP_013824546.1 | 3 - 4 | 666 | 76 | ||
GenBank mRNA | JT677480.1 | - | 2 - 3 | 425 | 76 | |
JT693924.1 | - | 4 - 5 | 571 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |