Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677916.3 | XP_005677973.2 | 2 - 3 | 2209 | 126 | |
XM_013962546.2 | XP_013818000.2 | 2 - 3 | 1569 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |