Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684239.2 | XP_005684296.2 | 7 - 8 | 1145 | 59 | |
XM_018052528.1 | XP_017908017.1 | 7 - 8 | 1178 | 59 | ||
XM_018052529.1 | XP_017908018.1 | 7 - 8 | 1137 | 59 | ||
GenBank mRNA | JO616831.2 | - | 1 - 2 | 509 | 59 | |
KA321793.1 | - | 1 - 2 | 742 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |