Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684767.3 | XP_005684824.2 | 4 - 5 | 846 | 68 | |
XM_005684768.3 | XP_005684825.2 | 4 - 5 | 846 | 68 | ||
GenBank mRNA | JO421239.2 | - | 4 - 5 | 869 | 68 | |
JO430014.2 | - | 4 - 5 | 820 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |