Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H11orf88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689428.2 | XP_005689485.2 | 2 - 3 | 585 | 87 | |
XM_018058999.1 | XP_017914488.1 | 3 - 4 | 736 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |