Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059672.1 | XP_017915161.1 | 6 - 7 | 1827 | 56 | |
XM_018059673.1 | XP_017915162.1 | 5 - 6 | 1678 | 56 | ||
GenBank mRNA | JO324077.2 | - | 5 - 6 | 1569 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |