Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692728.3 | XP_005692785.1 | 3 - 4 | 335 | 85 | |
XM_005692729.3 | XP_005692786.1 | 3 - 4 | 354 | 85 | ||
GenBank mRNA | JO404286.2 | - | 3 - 4 | 432 | 85 | |
JO441929.2 | - | 3 - 4 | 386 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |