Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692728.3 | XP_005692785.1 | 6 - 7 | 803 | 88 | |
XM_005692729.3 | XP_005692786.1 | 6 - 7 | 822 | 88 | ||
GenBank mRNA | JO404286.2 | - | 6 - 7 | 900 | 88 | |
JO441929.2 | - | 6 - 7 | 854 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |