Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051679.1 | XP_017907168.1 | 11 - 12 | 898 | 97 | |
GenBank mRNA | JO586435.2 | - | 1 - 2 | 661 | 97 | |
JO586437.2 | - | 1 - 2 | 661 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |