Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051679.1 | XP_017907168.1 | 2 - 3 | 173 | 65 | |
GenBank mRNA | JO586437.2 | - | 10 - 11 | 1365 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |