Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051903.1 | XP_017907392.1 | 3 - 4 | 285 | 80 | |
GenBank mRNA | JO429878.2 | - | 5 - 6 | 1787 | 80 | |
JO595714.2 | - | 5 - 6 | 1804 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |