Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056205.1 | XP_017911694.1 | 5 - 6 | 621 | 51 | |
XM_018056206.1 | XP_017911695.1 | 5 - 6 | 588 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |