Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039284.1 | XP_017894773.1 | 4 - 5 | 725 | 67 | |
XM_018039285.1 | XP_017894774.1 | 4 - 5 | 729 | 67 | ||
XM_018039286.1 | XP_017894775.1 | 4 - 5 | 743 | 67 | ||
XM_018039287.1 | XP_017894776.1 | 4 - 5 | 660 | 67 | ||
XM_018039288.1 | XP_017894777.1 | 4 - 5 | 596 | 67 | ||
GenBank mRNA | KA314265.1 | - | 4 - 5 | 652 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |